(2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(((3S,4R,4aS)-8-oxo-4-vinyl-3,4,4a,5,6,8-hexahydropyrano[3,4-c]pyran-3-yl)oxy)tetrahydro-2H-pyran-3-yl 3,3',5-trihydroxy-[1,1'-biphenyl]-2-carboxylate 97%

BLDPBD113763-100MGEA 228 EUR
BLDPBD113763-100MG
(2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(((3S,4R,4aS)-8-oxo-4-vinyl-3,4,4a,5,6,8-hexahydropyrano[3,4-c]pyran-3-yl)oxy)tetrahydro-2H-pyran-3-yl 3,3',5-trihydroxy-[1,1'-biphenyl]-2-carboxylate 97%
(2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(((3S,4R,4aS)-8-oxo-4-vinyl-3,4,4a,5,6,8-hexahydropyrano[3,4-c]pyran-3-yl)oxy)tetrahydro-2H-pyran-3-yl 3,3',5-trihydroxy-[1,1'-biphenyl]-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₉H₃₀O₁₃
MW: 586,5407 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD06656289
Núm. CAS: 21018-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR