(E)-Ethyl 2-cyano-3-phenylacrylate 95%

BLDPBD113408-5GEA 70 EUR
BLDPBD113408-5G BLDPBD113408-25G BLDPBD113408-1G
(E)-Ethyl 2-cyano-3-phenylacrylate 95%
(E)-Ethyl 2-cyano-3-phenylacrylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₁NO₂
MW: 201,2212 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00009137
Núm. CAS: 2169-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR