4-Iodo-1,2-dimethoxybenzene 97%

BLDPBD11186-1GEA 10 EUR
BLDPBD11186-1G BLDPBD11186-500G BLDPBD11186-10G BLDPBD11186-5G BLDPBD11186-100G BLDPBD11186-25G
4-Iodo-1,2-dimethoxybenzene 97%
4-Iodo-1,2-dimethoxybenzene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₉IO₂
MW: 264,0603 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00094416
Núm. CAS: 5460-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR