8-Nitroimidazo[1,2-a]pyridine 98%

BLDPBD111446-25GEA 665 EUR
BLDPBD111446-25G BLDPBD111446-250MG BLDPBD111446-0.025 BLDPBD111446-5G BLDPBD111446-1G
8-Nitroimidazo[1,2-a]pyridine 98%
8-Nitroimidazo[1,2-a]pyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₅N₃O₂
MW: 163,1335 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08056674
Núm. CAS: 52310-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR