8-Fluoro-5-nitroquinoline 98%

BLDPBD111286-1GEA 168 EUR
BLDPBD111286-1G BLDPBD111286-100MG BLDPBD111286-250MG BLDPBD111286-5G
8-Fluoro-5-nitroquinoline 98%
8-Fluoro-5-nitroquinoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₅FN₂O₂
MW: 192,1466 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09048716
Núm. CAS: 94832-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR