Ácido (R)-(-)-2-hidroxi-2-(2-clorofenil)acetico 98%
|
|
BLDPBD11030-100GEA
58
EUR
BLDPBD11030-100G
BLDPBD11030-10G
BLDPBD11030-500G
BLDPBD11030-5G
BLDPBD11030-25G
Ácido (R)-(-)-2-hidroxi-2-(2-clorofenil)acetico 98%
Ácido (R)-(-)-2-hidroxi-2-(2-clorofenil)acetico
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₇ClO₃ MW: 186,59 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00798429 Núm. CAS: 52950-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...