6-Thioguanosine 95%

BLDPBD110027-100MGEA 178 EUR
BLDPBD110027-100MG BLDPBD110027-1G BLDPBD110027-250MG
6-Thioguanosine 95%
6-Thioguanosine

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formula: C₁₀H₁₃N₅O₄S
MW: 299,30 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00005736
Núm. CAS: 85-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR