Methyl 6-methylbenzo[b]thiophene-2-carboxylate 98%

BLDPBD109719-250MGEA 86 EUR
BLDPBD109719-250MG BLDPBD109719-1G BLDPBD109719-100MG
Methyl 6-methylbenzo[b]thiophene-2-carboxylate 98%
Methyl 6-methylbenzo[b]thiophene-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₀O₂S
MW: 206,2600 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11044570
Núm. CAS: 82787-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR