6-Methyl-4-(trifluoromethyl)nicotinonitrile 95%

BLDPBD109696-250MGEA 37 EUR
BLDPBD109696-250MG BLDPBD109696-100MG BLDPBD109696-1G
6-Methyl-4-(trifluoromethyl)nicotinonitrile 95%
6-Methyl-4-(trifluoromethyl)nicotinonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₅F₃N₂
MW: 186,1300 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01570786
Núm. CAS: 13600-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR