(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol 95%

BLDPBD109430-250MGEA 55 EUR
BLDPBD109430-250MG BLDPBD109430-1G BLDPBD109430-100MG
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol 95%
(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₀H₁₂N₄O₄S
MW: 284,2917 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00005743
Núm. CAS: 574-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR