Isoquinoline-6-carbonitrile 97%

BLDPBD109411-5GEA 418 EUR
BLDPBD109411-5G BLDPBD109411-100MG BLDPBD109411-250MG BLDPBD109411-1G
Isoquinoline-6-carbonitrile 97%
Isoquinoline-6-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₆N₂
MW: 154,1680 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09991660
Núm. CAS: 106778-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR