6-Iodo-3,4-dihydronaphthalen-1(2H)-one 98%

BLDPBD109377-100MGEA 19 EUR
BLDPBD109377-100MG BLDPBD109377-5G BLDPBD109377-250MG BLDPBD109377-1G
6-Iodo-3,4-dihydronaphthalen-1(2H)-one 98%
6-Iodo-3,4-dihydronaphthalen-1(2H)-one

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₉IO
MW: 272,0800 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09751536
Núm. CAS: 340825-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR