6-Iodo-[1,2,4]triazolo[1,5-a]pyridine 97%

BLDPBD109373-250MGEA 21 EUR
BLDPBD109373-250MG BLDPBD109373-1G BLDPBD109373-25G BLDPBD109373-5G
6-Iodo-[1,2,4]triazolo[1,5-a]pyridine 97%
6-Iodo-[1,2,4]triazolo[1,5-a]pyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₄IN₃
MW: 245,021 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06797552
Núm. CAS: 614750-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR