5-Chloro-6-hydroxybenzo[d]oxazol-2(3H)-one 95+%

BLDPBD109338-250MGEA 324 EUR
BLDPBD109338-250MG BLDPBD109338-100MG BLDPBD109338-1G BLDPBD109338-5G
5-Chloro-6-hydroxybenzo[d]oxazol-2(3H)-one 95+%
5-Chloro-6-hydroxybenzo[d]oxazol-2(3H)-one

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₄ClNO₃
MW: 185,5700 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00143502
Núm. CAS: 1750-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR