2-(Methylamino)-1H-purin-6(7H)-one 98%

BLDPBD109306-1GEA 166 EUR
BLDPBD109306-1G BLDPBD109306-250MG BLDPBD109306-100MG
2-(Methylamino)-1H-purin-6(7H)-one 98%
2-(Methylamino)-1H-purin-6(7H)-one

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₇N₅O
MW: 165,15 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00127896
Núm. CAS: 10030-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR