4-Bromo-2-fluoro-5-methylbenzaldehyde 98%

BLDPBD10927-100MGEA 46 EUR
BLDPBD10927-100MG BLDPBD10927-250MG BLDPBD10927-1G
4-Bromo-2-fluoro-5-methylbenzaldehyde 98%
4-Bromo-2-fluoro-5-methylbenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₆BrFO
MW: 217,0400 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07782048
Núm. CAS: 916792-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR