6-Chloroimidazo[1,2-b]pyridazine 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD108932-100GEA 144 EUR
BLDPBD108932-0.025 BLDPBD108932-100G BLDPBD108932-5G BLDPBD108932-25G BLDPBD108932-1G BLDPBD108932-500G BLDPBD108932-10G
6-Chloroimidazo[1,2-b]pyridazine 97%
6-Chloroimidazo[1,2-b]pyridazine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₄ClN₃
MW: 153,5691 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07778345
Núm. CAS: 6775-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR