6-Chloro-2,2-difluorobenzo[d][1,3]dioxol-5-amine 97%
|
|
BLDPBD108684-1GEA
109
EUR
BLDPBD108684-1G
BLDPBD108684-250MG
BLDPBD108684-5G
6-Chloro-2,2-difluorobenzo[d][1,3]dioxol-5-amine 97%
6-Chloro-2,2-difluorobenzo[d][1,3]dioxol-5-amine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₇H₄ClF₂NO₂ MW: 207,56 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04972803 Núm. CAS: 73051-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...