5-Chlorobenzimidazole 97%
|
|
BLDPBD108648-25GEA
133
EUR
BLDPBD108648-25G
BLDPBD108648-1G
BLDPBD108648-100G
BLDPBD108648-250MG
BLDPBD108648-10G
BLDPBD108648-5G
5-Chlorobenzimidazole 97%
5-Chlorobenzimidazole
Storage condition: Sealed in dry, Room Temperature
Formula:
C₇H₅ClN₂ MW: 152,5810 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD07432850 Núm. CAS: 4887-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...