5-Methyl-2-thiophenecarbonitrile 95%

BLDPBD107343-25GEA 227 EUR
BLDPBD107343-25G BLDPBD107343-10G BLDPBD107343-1G BLDPBD107343-5G
5-Methyl-2-thiophenecarbonitrile 95%
5-Methyl-2-thiophenecarbonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₅NS
MW: 123,1756 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01434334
Núm. CAS: 72835-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR