Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate 98%

BLDPBD106052-1000GEA 25 EUR
BLDPBD106052-1000G BLDPBD106052-1G BLDPBD106052-500G BLDPBD106052-100G
Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate 98%
Zinc(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₂₂O₁₄Zn
MW: 455,70 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04037230
Núm. CAS: 4468-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR