(Z)-Ethyl 2-(2-aminothiazol-4-yl)-2-(hydroxyimino)acetate 97%

BLDPBD10594-10GEA 19 EUR
BLDPBD10594-10G BLDPBD10594-25G BLDPBD10594-5G
(Z)-Ethyl 2-(2-aminothiazol-4-yl)-2-(hydroxyimino)acetate 97%
(Z)-Ethyl 2-(2-aminothiazol-4-yl)-2-(hydroxyimino)acetate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₉N₃O₃S
MW: 215,22 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010415
Núm. CAS: 64485-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR