Benzyl glycidyl ether 98%

BLDPBD105469-5GEA 9 EUR
BLDPBD105469-5G BLDPBD105469-25G BLDPBD105469-10G
Benzyl glycidyl ether 98%
Benzyl glycidyl ether

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₂O₂
MW: 164,2011 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00068664
Núm. CAS: 2930-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR