5-Nitrobenzoxazole 98%

BLDPBD104962-1GEA 17 EUR
BLDPBD104962-1G BLDPBD104962-10G BLDPBD104962-25G BLDPBD104962-5G BLDPBD104962-100G
5-Nitrobenzoxazole 98%
5-Nitrobenzoxazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₄N₂O₃
MW: 164,1183 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD01359855
Núm. CAS: 70886-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR