5-Methylpyrazine-2-carboxamide 98%

BLDPBD104899-250MGEA 15 EUR
BLDPBD104899-250MG BLDPBD104899-5G BLDPBD104899-25G BLDPBD104899-1G
5-Methylpyrazine-2-carboxamide 98%
5-Methylpyrazine-2-carboxamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₇N₃O
MW: 137,1400 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00190581
Núm. CAS: 5521-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR