Ethyl 5-methyl-1H-indazole-3-carboxylate 96%

BLDPBD104658-1GEA 288 EUR
BLDPBD104658-1G BLDPBD104658-250MG BLDPBD104658-100MG
Ethyl 5-methyl-1H-indazole-3-carboxylate 96%
Ethyl 5-methyl-1H-indazole-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂N₂O₂
MW: 204,23 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06659781
Núm. CAS: 1908-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR