5-Mercapto-1-methyl-1H-tetrazole 98%

BLDPBD104419-5GEA 7 EUR
BLDPBD104419-5G BLDPBD104419-25G
5-Mercapto-1-methyl-1H-tetrazole 98%
5-Mercapto-1-methyl-1H-tetrazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂H₄N₄S
MW: 116,14 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00037317
Núm. CAS: 13183-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR