(2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid 95%
|
|
BLDPBD1036-100GEA
62
EUR
BLDPBD1036-100G
BLDPBD1036-25G
BLDPBD1036-10G
BLDPBD1036-5G
BLDPBD1036-500G
(2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid 95%
Fmoc-Thr-OH
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₁₉H₁₉NO₅ MW: 341,358 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00077072 Núm. CAS: 73731-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...