5-Chloro-3-iodo-2-methylaniline 98%

BLDPBD103509-100MGEA 33 EUR
BLDPBD103509-100MG BLDPBD103509-1G BLDPBD103509-250MG
5-Chloro-3-iodo-2-methylaniline 98%
5-Chloro-3-iodo-2-methylaniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₇ClIN
MW: 267,5000 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD08690116
Núm. CAS: 870606-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR