2,4-Dichloro-5-sulphamoylbenzoic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD10341-10GEA 7 EUR
BLDPBD10341-10G BLDPBD10341-100G BLDPBD10341-25G BLDPBD10341-500G
2,4-Dichloro-5-sulphamoylbenzoic acid 98%
2,4-Dichloro-5-sulphamoylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₅Cl₂NO₄S
MW: 270,09 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00007931
Núm. CAS: 2736-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR