5-Chloro-2,2-difluoro-1,3-benzodioxole 98%

BLDPBD103298-250MGEA 23 EUR
BLDPBD103298-250MG BLDPBD103298-5G BLDPBD103298-1G
5-Chloro-2,2-difluoro-1,3-benzodioxole 98%
5-Chloro-2,2-difluoro-1,3-benzodioxole

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₃ClF₂O₂
MW: 192,54 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD04972706
Núm. CAS: 72769-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR