5-Chloro-1H-indazole 98%

BLDPBD103184-25GEA 379 EUR
BLDPBD103184-25G BLDPBD103184-5G BLDPBD103184-100MG BLDPBD103184-10G BLDPBD103184-250MG BLDPBD103184-100G BLDPBD103184-1G
5-Chloro-1H-indazole 98%
5-Chloro-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₅ClN₂
MW: 152,5800 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00022785
Núm. CAS: 698-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR