2-Mercapto-6-nitrobenzothiazole 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD10314-1GEA 37 EUR
BLDPBD10314-1G
2-Mercapto-6-nitrobenzothiazole 97%
2-Mercapto-6-nitrobenzothiazole
Formula: C₇H₄N₂O₂S₂
MW: 212,24 g/mol
Número MDL: MFCD00041850
Núm. CAS: 4845-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR