5-Bromoimidazo[1,2-a]pyridine 96%
|
|
BLDPBD103048-10GEA
167
EUR
BLDPBD103048-10G
BLDPBD103048-5G
BLDPBD103048-25G
BLDPBD103048-1G
BLDPBD103048-250MG
5-Bromoimidazo[1,2-a]pyridine 96%
5-Bromoimidazo[1,2-a]pyridine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₅BrN₂ MW: 197,03 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08275698 Núm. CAS: 69214-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...