5-Bromo-6-fluoro-1H-indazole 97%

BLDPBD103003-25GEA 1270 EUR
BLDPBD103003-25G BLDPBD103003-0.025 BLDPBD103003-250MG BLDPBD103003-1G BLDPBD103003-5G
5-Bromo-6-fluoro-1H-indazole 97%
5-Bromo-6-fluoro-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₄BrFN₂
MW: 215,0225 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11044589
Núm. CAS: 105391-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR