5-Bromo-3-iodo-1H-indazole 98%

BLDPBD102886-1GEA 10 EUR
BLDPBD102886-1G BLDPBD102886-10G BLDPBD102886-250MG BLDPBD102886-5G BLDPBD102886-25G BLDPBD102886-100G
5-Bromo-3-iodo-1H-indazole 98%
5-Bromo-3-iodo-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₄BrIN₂
MW: 322,9300 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07781637
Núm. CAS: 459133-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR