5-Bromo-3-fluorosalicylaldehyde 95%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD102873-1GEA
29
EUR
BLDPBD102873-1G
BLDPBD102873-25G
BLDPBD102873-250MG
BLDPBD102873-5G
5-Bromo-3-fluorosalicylaldehyde 95%
5-Bromo-3-fluorosalicylaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₄BrFO₂ MW: 219,0079 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD03788530 Núm. CAS: 251300-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...