(5-Bromo-1,3-phenylene)dimethanol 95%

BLDPBD102529-100MGEA 49 EUR
BLDPBD102529-100MG BLDPBD102529-5G BLDPBD102529-250MG BLDPBD102529-1G
(5-Bromo-1,3-phenylene)dimethanol 95%
(5-Bromo-1,3-phenylene)dimethanol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₉BrO₂
MW: 217,0599 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00229250
Núm. CAS: 51760-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR