1,3,5-Triazine-2,4(1H,3H)-dione 95%

BLDPBD102418-1GEA 224 EUR
BLDPBD102418-1G BLDPBD102418-250MG BLDPBD102418-100MG
1,3,5-Triazine-2,4(1H,3H)-dione 95%
1,3,5-Triazine-2,4(1H,3H)-dione

Storage condition: Sealed in dry, Room Temperature

Formula: C₃H₃N₃O₂
MW: 113,0748 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00023157
Núm. CAS: 71-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR