5,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid 95%

BLDPBD101752-250MGEA 102 EUR
BLDPBD101752-250MG BLDPBD101752-5G BLDPBD101752-1G
5,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid 95%
5,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₉NO₃
MW: 167,16 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02642444
Núm. CAS: 51727-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR