5H-Cyclopenta[c]pyridin-7(6H)-one 95%
|
|
BLDPBD101680-100MGEA
106
EUR
BLDPBD101680-100MG
BLDPBD101680-250MG
BLDPBD101680-1G
5H-Cyclopenta[c]pyridin-7(6H)-one 95%
5H-Cyclopenta[c]pyridin-7(6H)-one
Storage condition: Sealed in dry, 2-8°C
Formula:
C₈H₇NO MW: 133,14 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD08694510 Núm. CAS: 51907-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...