5-(4-Bromophenyl)thiophene-2-carbaldehyde 95%

BLDPBD100927-1GEA 207 EUR
BLDPBD100927-1G BLDPBD100927-250MG BLDPBD100927-100MG
5-(4-Bromophenyl)thiophene-2-carbaldehyde 95%
5-(4-Bromophenyl)thiophene-2-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₇BrOS
MW: 267,1417 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00829312
Núm. CAS: 38401-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR