5-(4-Bromophenyl)isothiazole 95%

BLDPBD100907-100MGEA 110 EUR
BLDPBD100907-100MG BLDPBD100907-1G BLDPBD100907-250MG
5-(4-Bromophenyl)isothiazole 95%
5-(4-Bromophenyl)isothiazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆BrNS
MW: 240,1200 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07779064
Núm. CAS: 49602-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR