5-(2-Thioxo-2,3-dihydrothiazol-4-yl)thiophene-2-carboxamide 95+%

BLDPBD100746-100GEA 738 EUR
BLDPBD100746-100G BLDPBD100746-25G BLDPBD100746-5G
5-(2-Thioxo-2,3-dihydrothiazol-4-yl)thiophene-2-carboxamide 95+%
5-(2-Thioxo-2,3-dihydrothiazol-4-yl)thiophene-2-carboxamide

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₈H₆N₂OS₃
MW: 242,3400 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD11040250
Núm. CAS: 52560-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR