6,7-Dihydro-5H-indeno[5,6-d][1,3]dioxol-5-one 95%

BLDPBD100539-5GEA 543 EUR
BLDPBD100539-5G BLDPBD100539-25G BLDPBD100539-250MG BLDPBD100539-1G BLDPBD100539-100MG
6,7-Dihydro-5H-indeno[5,6-d][1,3]dioxol-5-one 95%
6,7-Dihydro-5H-indeno[5,6-d][1,3]dioxol-5-one

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₈O₃
MW: 176,1687 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01925052
Núm. CAS: 6412-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR