3-(Oxiran-2-yl)-7-oxabicyclo[4.1.0]heptane 95%

BLDPBD100534-1GEA 22 EUR
BLDPBD100534-1G BLDPBD100534-5G BLDPBD100534-250MG BLDPBD100534-10G
3-(Oxiran-2-yl)-7-oxabicyclo[4.1.0]heptane 95%
3-(Oxiran-2-yl)-7-oxabicyclo[4.1.0]heptane

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₁₂O₂
MW: 140,1797 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00022354
Núm. CAS: 106-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR