4-tert-Butoxybenzonitrile 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD100262-5GEA 93 EUR
BLDPBD100262-5G BLDPBD100262-1G
4-tert-Butoxybenzonitrile 95%
4-tert-Butoxybenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₃NO
MW: 175,23 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03425273
Núm. CAS: 185259-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR