1,5-Dimethyl-1H-pyrazole-3-carboxylic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD10014-5GEA 22 EUR
BLDPBD10014-5G BLDPBD10014-100G BLDPBD10014-1G BLDPBD10014-10G BLDPBD10014-25G
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid 98%
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₈N₂O₂
MW: 140,14 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00085040
Núm. CAS: 5744-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR