(R)-(-)-1-Benzyl-3-hydroxypiperidine 95%
|
|
BLDPBD0890-5GEA
232
EUR
BLDPBD0890-5G
BLDPBD0890-1G
(R)-(-)-1-Benzyl-3-hydroxypiperidine 95%
(R)-(-)-1-Benzyl-3-hydroxypiperidine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₂H₁₇NO MW: 191,27 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00274307 Núm. CAS: 91599-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...