4-Amino-1-boc-piperidine 98%

BLDPBD0337-1GEA 6 EUR
BLDPBD0337-1G BLDPBD0337-0.5G BLDPBD0337-5G BLDPBD0337-25G BLDPBD0337-1000G BLDPBD0337-100G BLDPBD0337-10G BLDPBD0337-500G
4-Amino-1-boc-piperidine 98%
4-Amino-1-boc-piperidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01076201
Núm. CAS: 87120-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR